missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Triphenylacetic Acid, TRC

Product Code. 30012137
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012137 1 g 1g
30010192 500 mg 500mg
30007273 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012137 Supplier Toronto Research Chemicals Supplier No. TRCT8956951G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 595-91-5
Molecular Formula C20 H16 O2
Molecular Weight (g/mol) 288.34
Synonym Triphenylacetic acid, Benzeneacetic acid, α,α-diphenyl-, Acetic acid, triphenyl- (6CI,7CI,8CI), α,α-Diphenylbenzeneacetic acid, 2,2,2-Triphenylacetic acid, NSC 61, Triphenylacetic acid, Triphenylmethanecarboxylic acid, GI 179710X
IUPAC Name 2,2,2-triphenylacetic acid
SMILES OC(=O)C(c1ccccc1)(c2ccccc2)c3ccccc3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H16O2/c21-19(22)20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H,(H,21,22)
Formula Weight 288.115
Chemical Name or Material Triphenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.