missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Triphenylacetophenone, TRC

Product Code. 30456502
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456502 10 g 10g
30453587 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30456502 Supplier Toronto Research Chemicals Supplier No. TRCT80878510G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 466-37-5
Molecular Formula C26H20O
Molecular Weight (g/mol) 348.45
Synonym Benzopinacolone, NSC 194, NSC 2820, Phenyl trityl ketone, Tetraphenylethanone, ω,ω,ω-Triphenylacetophenone, beta-Benzopinacolone (6CI), 1,2,2,2-Tetraphenyl-1-ethanone, 1,2,2,2-Tetraphenylethanone, Tetraphenylethanone, 2,2,2-Triphenylacetophenone, 1,2,2,2-Tetraphenylethanone
IUPAC Name 1,2,2,2-tetraphenylethanone
SMILES O=C(c1ccccc1)C(c2ccccc2)(c3ccccc3)c4ccccc4

Specifications

Quantity 10 g
InChI Formula InChI=1S/C26H20O/c27-25(21-13-5-1-6-14-21)26(22-15-7-2-8-16-22,23-17-9-3-10-18-23)24-19-11-4-12-20-24/h1-20H
Formula Weight 348.1514
Chemical Name or Material 2,2,2-Triphenylacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.