missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3-Tetrafluoropropyl methacrylate, 97%, stab. with 50ppm 4-methoxyphenol, Thermo Scientific Chemicals

Product Code. 11405327
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405327 5 g 5g
11415327 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11405327 Supplier Thermo Scientific Alfa Aesar Supplier No. B22682.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 45102-52-1
Molecular Formula C7H8F4O2
Molecular Weight (g/mol) 200.133
MDL Number MFCD00042381
InChI Key RSVZYSKAPMBSMY-UHFFFAOYSA-N
Synonym 2,2,3,3-tetrafluoropropyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,3,3-tetrafluoropropyl ester, methacrylic acid 2,2,3,3-tetrafluoropropyl ester, 1h,1h,3h-tetrafluoropropyl methacrylate, 2,2,3,3-tetrafluoropropyl methacrylate, stab. with 2,2'-methylenebis 6-tert-butyl-4-methyl-phenol, acmc-1ajz7, ksc489s8d, rsvzyskapmbsmy-uhfffaoysa, 2,2,3,3-tetrafluoroprop-1-yl methacrylat
PubChem CID 123515
IUPAC Name 2,2,3,3-tetrafluoropropyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(C(F)F)(F)F

Specifications

Density 1.254
Boiling Point 146°C to 149°C
Flash Point 50°C (122°F)
Refractive Index 1.375
Quantity 5 g
UN Number UN3272
Beilstein 3588831
Formula Weight 200.13
Percent Purity 97%
Chemical Name or Material 2,2,3,3-Tetrafluoropropyl methacrylate, stabilized with 50ppm 4-methoxyphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.