missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,4-Tetrafluoro-1,3-dithietane, 97%

Product Code. 11437244
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437244 10 g 10g
11447244 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11437244 Supplier Thermo Scientific Alfa Aesar Supplier No. B21620.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1717-50-6 | C2F4S2 | 164.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1717-50-6
Molecular Formula C2F4S2
Molecular Weight (g/mol) 164.14
MDL Number MFCD00053538
InChI Key MPOCEMBRPHOCMK-UHFFFAOYSA-N
Synonym tetrafluoro-1,3-dithietane, pubchem3989, acmc-1budt, 1, 2,2,4,4-tetrafluoro, 2,4,4-tetrafluoro-1,3-dithietane, 1,3-dithietane,2,2,4,4-tetrafluoro, 2,2,4,4-tetrafluoro-1,3-dithietane
PubChem CID 74392
IUPAC Name 2,2,4,4-tetrafluoro-1,3-dithietane
SMILES FC1(F)SC(F)(F)S1

Specifications

Density 1.6
Boiling Point 47°C to 49°C
Flash Point 36°C (96°F)
Odor Unpleasant
Refractive Index 1.393
Quantity 10 g
UN Number UN1993
Formula Weight 164.14
Percent Purity 97%
Chemical Name or Material 2,2,4,4-Tetrafluoro-1,3-dithietane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.