missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,4-Tetramethylcyclohexanecarboxylic Acid, TRC

Product Code. 30457031
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457031 25 mg 25mg
30457203 10 mg 10mg
30453316 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457031 Supplier Toronto Research Chemicals Supplier No. TRCT82425525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2,4,4-Tetramethylcyclohexanecarboxylic Acid
Analyte or Component Names 2,2,4,4-Tetramethylcyclohexanecarboxylic Acid
Shipping Condition Room Temperature
Molecular Formula C11 H20 O2
InChI Formula InChI=1S/C11H20O2/c1-10(2)6-5-8(9(12)13)11(3,4)7-10/h8H,5-7H2,1-4H3,(H,12,13)
Solution Type Neat
SMILES CC1(C)CCC(C(=O)O)C(C)(C)C1
IUPAC Name 2,2,4,4-tetramethylcyclohexane-1-carboxylic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 184.275
Quantity 25 mg
Formula Weight 184.146
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.