missing translation for 'onlineSavingsMsg'
Learn More

2,2',6,6'-Tetra-tert-butylbisphenol F, Dr. Ehrenstorfer

Product Code. 30318786
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318786 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318786 Supplier Dr. Ehrenstorfer Supplier No. DREC17428900
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2',6,6'-Tetra-tert-Butylbisphenol F
CAS 118-82-1
Analyte or Component Names 2,2',6,6'-Tetra-tert-Butylbisphenol F
Shipping Condition +20°C
Molecular Formula C29 H44 O2
Certifications/Compliance ISO/IEC 17025
Synonym 4,4'-Methylenebis[2,6-bis(1,1-dimethylethyl)phenol (ACI), Phenol, 4,4'-methylenebis[2,6-di-tert-butyl- (6CI, 8CI), 2,6-Di-tert-butyl-4-[(3,5-di-tert-butyl-4-hydroxyphenyl)methyl]phenol, 2,6-Ditert-butyl-4-[(3,5-ditert-butyl-4-hydroxyphenyl)methyl]phenol, 3,3',5,5'-Tetra-tert-butyl-4,4'-dihydroxydiphenylmethane, 4,4'-Methylene-bis-2,6-tert-butylphenol, 4,4'-Methylenebis[2,6-di-tert-butylphenol], Agidol 23, Antioxidant 4426, Antioxidant 702, Antioxidant 728, Antioxidant E 702, Antioxidant T 511, AO 4426, Binox M, Bis(3,5-di-tert-butyl-4-hydroxyphenyl)methane, Bis(4-hydroxy-3,5-di-tert-butylphenyl)methane, Di(3,5-di-tert-butyl-4-hydroxyphenyl)methane;2,2',6,6'-Tetra-tert-Butylbisphenol F
InChI Formula InChI=1S/C29H44O2/c1-26(2,3)20-14-18(15-21(24(20)30)27(4,5)6)13-19-16-22(28(7,8)9)25(31)23(17-19)29(10,11)12/h14-17,30-31H,13H2,1-12H3
SMILES CC(C)(C)c1cc(Cc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)cc(c1O)C(C)(C)C
IUPAC Name 2,6-ditert-butyl-4-[(3,5-ditert-butyl-4-hydroxyphenyl)methyl]phenol
Molecular Weight (g/mol) 424.659
Quantity 10 mg
Formula Weight 424.334 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.