missing translation for 'onlineSavingsMsg'
Learn More

(2,2,6,6-Tetramethyl-3,5-heptanedionato)lithium, 98%, Thermo Scientific Chemicals

Product Code. 11368907
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368907 5 g 5g
11378907 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368907 Supplier Thermo Scientific Alfa Aesar Supplier No. 038596.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used as reactant for synthesis of Bis-cycloiridiated complexes for electroluminescent studies, cationic imido alkylidene complexes, polyaromatic ligands containing pyrene and mono-cyclometalated platinum complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22441-13-0
MDL Number MFCD00142754
Synonym Lithium 2,2,6,6-tetramethyl-3,5-heptanedionate; Li(TMHD)

Specifications

Melting Point 127°C
Boiling Point 180°C (0.05 mmHg)
Quantity 5 g
Solubility Information Soluble in acetone and methanol.
Formula Weight 190.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (2,2,6,6-Tetramethyl-3,5-heptanedionato)lithium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.