missing translation for 'onlineSavingsMsg'
Learn More

2-(2,6-Dichlorophenyl)ethanimidamide hydrochloride, 97%, Thermo Scientific™

Product Code. 10203292
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10203292 1 g Amber Glass Bottle 1g
10709225 5 g Amber Glass Bottle 5g
10355062 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10203292 Supplier Thermo Scientific Maybridge Supplier No. SPB00311DA
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 175276-76-3
Molecular Formula C8H9Cl3N2
Molecular Weight (g/mol) 239.524
MDL Number MFCD00053014
InChI Key YNPZJFKKDYCULJ-UHFFFAOYSA-N
Synonym 2-2,6-dichlorophenyl ethanimidamide hydrochloride, 2-2,6-dichlorophenyl acetimidamide hydrochloride, 2,6-dichlorophenylacetamidinehydrochloride, 2,6-dichlorophenylacetamidine hydrochloride, 2,6-dichlorobenzeneethanimidamide hcl, 2,6-dichlorophenylacetamidine hydrochlorid, 2-2,6-dichlorophenyl ethanamidine, chloride, 2-2,6-dichlorophenyl ethaneimidamide hydrochloride, 2-2,6-bis chloranyl phenyl ethanimidamide hydrochloride, benzeneethanimidamide,2,6-dichloro-, hydrochloride 1:1
PubChem CID 12690064
IUPAC Name 2-(2,6-dichlorophenyl)ethanimidamide;hydrochloride
SMILES C1=CC(=C(C(=C1)Cl)CC(=N)N)Cl.Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3CH2C(=NH)NH2·HCl
Quantity 1 g
Formula Weight 239.53
Percent Purity 97%
Chemical Name or Material 2-(2, 6-Dichlorophenyl)ethanimidamide hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.