missing translation for 'onlineSavingsMsg'
Learn More

2,2,6-Trimethyl-1,3-dioxin-4-one, cont. up to ca 6% acetone

Product Code. 11442387
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11442387

Brand: Thermo Scientific Alfa Aesar A14254.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 5394-63-8 | C7H10O3 | 142.154 g/mol

2,2,6-Trimethyl-1,3-dioxin-4-one is used in the synthesis of dihydropyrimininones as picomolar sodium iodide symporter inhibitors. It is also used in the synthesis of resorcinol amide Hsp90 Inhibitor AT13387. It is used in the synthesis of acetylketene by flash pyrolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,6-Trimethyl-1,3-dioxin-4-one is used in the synthesis of dihydropyrimininones as picomolar sodium iodide symporter inhibitors. It is also used in the synthesis of resorcinol amide Hsp90 Inhibitor AT13387. It is used in the synthesis of acetylketene by flash pyrolysis.

Solubility
Soluble in Chloroform, Methanol. Not miscible in water.

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5394-63-8
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD00040468
InChI Key XFRBXZCBOYNMJP-UHFFFAOYSA-N
Synonym 2,2,6-trimethyl-4h-1,3-dioxin-4-one, 2,2,4-trimethyl-6-keto-1,3-dioxin, diketene acetone adduct, 4h-1,3-dioxin-4-one, 2,2,6-trimethyl, 2,2,6-trimethyl-1,3-dioxen-4-one, 2,2,6-trimethyl-1,3-dioxine-4-one, 2,2,6-trimethyl-1,3 dioxin-4-one, pubchem20364, 2,2,6-trimethyl-4h-1,3-dioxin-4-on, acmc-1akcb
PubChem CID 79368
IUPAC Name 2,2,6-trimethyl-1,3-dioxin-4-one
SMILES CC1=CC(=O)OC(O1)(C)C

Specifications

Melting Point 7°C
Density 1.094
Boiling Point 65°C to 67°C (2 mmHg)
Flash Point 14°C (57°F)
Refractive Index 1.461
Quantity 100 g
UN Number UN1993
Beilstein 2408
Solubility Information Soluble in Chloroform,Methanol. Not miscible in water.
Formula Weight 142.15
Percent Purity 94%
Assay cont. up to ≈6% acetone
Chemical Name or Material 2,2,6-Trimethyl-1,3-dioxin-4-one
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.