missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Biphenol, 99%

Product Code. 10656162 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10656162 100 g Plastic bottle 100g
10193340 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10656162 Supplier Thermo Scientific Acros Supplier No. 151991000
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1806-29-7 | C12H10O2 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1806-29-7
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00002210
InChI Key IMHDGJOMLMDPJN-UHFFFAOYSA-N
Synonym 2,2'-biphenol, 1,1'-biphenyl-2,2'-diol, 2,2'-dihydroxybiphenyl, biphenyl-2,2'-diol, o,o'-diphenol, o,o'-biphenol, o-dihydroxydiphenyl, o,o'-dihydroxybiphenyl, 2,2'-biphenyldiol, 2,2'-dihydroxydiphenyl
PubChem CID 15731
ChEBI CHEBI:28970
IUPAC Name 2-(2-hydroxyphenyl)phenol
SMILES OC1=CC=CC=C1C1=CC=CC=C1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 108°C to 111°C
Color Beige to Beige-Gray
Density 0.8300g/mL
Boiling Point 315°C
Flash Point 154°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula HOC6H4C6H4OH
Quantity 100 g
Beilstein 06, 989
Specific Gravity 0.83
Solubility Information Solubility in water: insoluble
Formula Weight 186.21
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2,2′-Biphenol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2,2'-Biphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.