missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis[4-(4-aminophenoxy)phenyl]propane, 98%

Product Code. 15481178
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15481178 100 g 100g
15471178 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15481178 Supplier Thermo Scientific Alfa Aesar Supplier No. H55057.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13080-86-9 | C27H26N2O2 | 410.517 g/mol

It is applied in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in pharmaceutical industry.

Solubility
Very slightly soluble in water. Soluble in acetone, Dimethyl sulfoxide(DMSO).

Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13080-86-9
Molecular Formula C27H26N2O2
Molecular Weight (g/mol) 410.517
MDL Number MFCD00039152
InChI Key KMKWGXGSGPYISJ-UHFFFAOYSA-N
Synonym 2,2-bis 4-4-aminophenoxy phenyl propane, 4,4'-propane-2,2-diylbis 4,1-phenylene bis oxy dianiline, 2,2'-bis 4-aminophenoxyphenyl propane, benzenamine, 4,4'-1-methylethylidene bis 4,1-phenyleneoxy bis, 4,4'-isopropylidenebis 4,1-phenyleneoxy dianiline, 4,4'-4,4'-isopropylidenediphenyl-1,1'-diyldioxy dianiline, bis 4-4-aminophenoxy phenyl dimethyl methane, 4,4'-1-methylethylidene bis 4,1-phenyleneoxy bisbenzenamine
PubChem CID 83119
IUPAC Name 4-[4-[2-[4-(4-aminophenoxy)phenyl]propan-2-yl]phenoxy]aniline
SMILES CC(C)(C1=CC=C(C=C1)OC2=CC=C(C=C2)N)C3=CC=C(C=C3)OC4=CC=C(C=C4)N

Specifications

Melting Point 127°C to 130°C
Quantity 100 g
Solubility Information Very slightly soluble in water. Soluble in acetone,Dimethyl sulfoxide(DMSO).
Formula Weight 410.52
Percent Purity 98%
Chemical Name or Material 2,2-Bis[4-(4-aminophenoxy)phenyl]propane
compliance-icons
Product Identifier
  • 2,2-Bis[4-(4-aminophenoxy)phenyl]propane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P330-Rinse mouth.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 4,4'-[(1-methylethylidene)bis(4,1-phenyleneoxy)]bisbenzenamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.