missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bis(trifluoromethyl)benzidine, 97%

Product Code. 15595976 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15595976 25 g 25g
15575976 1 g 1g
15585976 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15595976 Supplier Thermo Scientific Acros Supplier No. 457570250
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 341-58-2 | C14H10F6N2 | 320.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 341-58-2
Molecular Formula C14H10F6N2
Molecular Weight (g/mol) 320.24
MDL Number MFCD00190155
InChI Key NVKGJHAQGWCWDI-UHFFFAOYSA-N
Synonym 2,2'-bis trifluoromethyl benzidine, 2,2'-bis trifluoromethyl-1,1'-biphenyl-4,4'-diamine, 4-4-amino-2-trifluoromethyl phenyl-3-trifluoromethyl aniline, 2,2'-bis trifluoromethyl-4,4'-diaminobiphenyl, 2,2'-bis-trifluoromethyl-biphenyl-4,4'-diamine, tfmb, 2,2'-bis trifluoromethyl-4,4'-biphenyldiamine, 4,4'-diamino-2,2'-bis trifluoromethyl biphenyl, 1,1'-biphenyl-4,4'-diamine, 2,2'-bis trifluoromethyl, 4-4-azanyl-2-trifluoromethyl phenyl-3-trifluoromethyl aniline
PubChem CID 629349
IUPAC Name 4-[4-amino-2-(trifluoromethyl)phenyl]-3-(trifluoromethyl)aniline
SMILES NC1=CC(=C(C=C1)C1=C(C=C(N)C=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 183.0°C to 184.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 25 g
Formula Weight 320.24
Percent Purity 97%
Chemical Name or Material 2,2'-Bis(trifluoromethyl)benzidine
compliance-icons
Product Identifier
  • 2,2'-Bis(trifluoromethyl)benzidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.