missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(Bromomethyl)phenyl]thiophene, ≥95%, Thermo Scientific™

Product Code. 10279222
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10279222 1 g 1g
10035852 250 mg 250mg
10112804 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10279222 Supplier Thermo Scientific Maybridge Supplier No. CC35508DA
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 791078-04-1
Molecular Formula C11H9BrS
Molecular Weight (g/mol) 253.157
MDL Number MFCD07368537
InChI Key IJNVNLWIQXMBPA-UHFFFAOYSA-N
PubChem CID 7060555
IUPAC Name 2-[2-(bromomethyl)phenyl]thiophene
SMILES C1=CC=C(C(=C1)CBr)C2=CC=CS2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Synonym 2-2-bromomethyl phenyl thiophene, 2-thien-2-yl benzyl bromide, thiophene,2-2-bromomethyl phenyl, 1-bromomethyl-2-thien-2-yl benzene, thiophene, 2-2-bromomethyl phenyl
Formula Weight 253.16
Percent Purity 95%
Chemical Name or Material 2-[2-(Bromomethyl)phenyl]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.