missing translation for 'onlineSavingsMsg'
Learn More

2-(2-chlorophenoxy)ethylamine, 97%, Thermo Scientific™

Product Code. 10181032
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10181032 250 mg Amber Glass Bottle 250mg
10141082 1 g Amber Glass Bottle 1g
10595364 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10181032 Supplier Thermo Scientific Maybridge Supplier No. RF03300CB
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 26378-53-0
Molecular Formula C8H10ClNO
Molecular Weight (g/mol) 171.624
MDL Number MFCD00125294
InChI Key NAPNYPMMDVRKGM-UHFFFAOYSA-N
PubChem CID 2735788
IUPAC Name 2-(2-chlorophenoxy)ethanamine
SMILES C1=CC=C(C(=C1)OCCN)Cl

Specifications

CAS Min % 97.0
Quantity 250 mg
Packaging Amber Glass Bottle
Assay Percent Range 97%
Linear Formula ClC6H4O(CH2)2NH2
Synonym 2-2-chlorophenoxy ethylamine, 2-2-chlorophenoxy ethanamine, 2-2-chlorophenoxy-1-ethanamine, ethanamine, 2-2-chlorophenoxy, 2-2-chlorophenoxy ethyl amine, 1-2-aminoethoxy-2-chlorobenzene, acmc-20aoqz, ethanamine,2-2-chlorophenoxy, 2-2-chlorophenoxy ethan-1-amine
Formula Weight 171.63
Percent Purity 97%
Chemical Name or Material 2-(2-Chlorophenoxy)ethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.