missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Chlorophenoxy)propionic acid, Dr. Ehrenstorfer

Product Code. 30311864
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30311864 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311864 Supplier Dr. Ehrenstorfer Supplier No. DREC11557500
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2-Chlorophenoxy)propanoic acid
CAS 25140-86-7
Analyte or Component Names 2-(2-Chlorophenoxy)propanoic acid
Shipping Condition +20°C
Molecular Formula C9 H9 Cl O3
Certifications/Compliance ISO/IEC 17025
Synonym Propionic acid, 2-(o-chlorophenoxy)- (6CI,7CI,8CI), (RS)-2-(2-Chlorophenoxy)propionic acid, (±)-2-(2-Chlorophenoxy)propanoic acid, (±)-2-(2-Chlorophenoxy)propionic acid, 2-(2-Chlorophenoxy)propionic acid, 2-(o-Chlorophenoxy)propionic acid
InChI Formula InChI=1S/C9H9ClO3/c1-6(9(11)12)13-8-5-3-2-4-7(8)10/h2-6H,1H3,(H,11,12)
SMILES CC(Oc1ccccc1Cl)C(=O)O
IUPAC Name 2-(2-chlorophenoxy)propanoic acid
Molecular Weight (g/mol) 200.619
Quantity 100 mg
Formula Weight 200.024 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.