missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dichlorobenzophenone, 98+%

Product Code. 11352396
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352396 5 g 5g
11342396 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352396 Supplier Thermo Scientific Alfa Aesar Supplier No. L06144.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5293-97-0 | C13H8Cl2O | 251.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5293-97-0
Molecular Formula C13H8Cl2O
Molecular Weight (g/mol) 251.106
MDL Number MFCD00039303
InChI Key DRDRZHJTTDSOPK-UHFFFAOYSA-N
Synonym 2,2'-dichlorobenzophenone, bis 2-chlorophenyl methanone, methanone, bis 2-chlorophenyl, unii-5y7f5u8ant, 5y7f5u8ant, benzophenone, 2,2'-dichloro, bis 2-chlorophenyl ketone, acmc-1ao0c, benzophenone,2'-dichloro, 2,2-dichlorobenzophenone
PubChem CID 347097
IUPAC Name bis(2-chlorophenyl)methanone
SMILES C1=CC=C(C(=C1)C(=O)C2=CC=CC=C2Cl)Cl

Specifications

Melting Point 50°C to 53°C
Quantity 5 g
Beilstein 2050630
Formula Weight 251.11
Percent Purity ≥98%
Chemical Name or Material 2,2'-Dichlorobenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.