missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dihydroxybiphenyl, 99%

Product Code. 11414130
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414130 25 g 25g
11492537 100 g 100g
11402547 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414130 Supplier Thermo Scientific Alfa Aesar Supplier No. A14800.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1806-29-7 | C12H10O2 | 186.21 g/mol

2,2'-Dihydroxybiphenyl, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Dihydroxybiphenyl, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1806-29-7
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00002210
InChI Key IMHDGJOMLMDPJN-UHFFFAOYSA-N
Synonym 2,2'-biphenol, 1,1'-biphenyl-2,2'-diol, 2,2'-dihydroxybiphenyl, biphenyl-2,2'-diol, o,o'-diphenol, o,o'-biphenol, o-dihydroxydiphenyl, o,o'-dihydroxybiphenyl, 2,2'-biphenyldiol, 2,2'-dihydroxydiphenyl
PubChem CID 15731
ChEBI CHEBI:28970
SMILES OC1=CC=CC=C1C1=CC=CC=C1O

Specifications

Melting Point 107°C to 111°C
Density 1.34
Boiling Point 314°C to 316°C
Flash Point 154°C (309°F)
Quantity 25 g
Beilstein 1638363
Solubility Information Insoluble in water.
IUPAC Name 2-(2-hydroxyphenyl)phenol
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 2,2'-Dihydroxybiphenyl
Show More Show Less
compliance-icons
Product Identifier
  • 2,2'-Dihydroxybiphenyl
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: [1,1-Biphenyl]-2,2-diol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.