missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethoxy-2-phenylacetophenone, 99%

Product Code. 10184010 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10184010 50 g Glass bottle 50g
10242652 5 g Glass bottle 5g
10543361 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10184010 Supplier Thermo Scientific Acros Supplier No. 187840500
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24650-42-8 | C16H16O3 | 256.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24650-42-8
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.30
MDL Number MFCD00008475
InChI Key KWVGIHKZDCUPEU-UHFFFAOYSA-N
Synonym 2,2-dimethoxy-2-phenylacetophenone, benzil dimethyl ketal, kayacure bdmk, irgacure 651, ethanone, 2,2-dimethoxy-1,2-diphenyl, lucirin bdk, photomer 51, benzil dimethylketal, esacure kb 1, irgacure 621
PubChem CID 90571
IUPAC Name 2,2-dimethoxy-1,2-diphenylethanone
SMILES COC(OC)(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 64°C to 68°C
Color White to Yellow
Loss on Drying 0.2% max. (75°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5COC(OCH3)2C6H5
Quantity 50 g
Merck Index 14, 3392
Formula Weight 256.29
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 2-Dimethoxy-2-phenylacetophenone, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.