missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diphenyl-1-picrylhydrazyl, TRC

Product Code. 30005423
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005423 1 g 1g
30009254 50 mg 50mg
30012433 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005423 Supplier Toronto Research Chemicals Supplier No. TRCD4920451G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1898-66-4
Molecular Formula C18 H12 N5 O6
Molecular Weight (g/mol) 394.32
Synonym 2,2-Diphenyl-1-picrylhydrazyl (DPPH Radical)
IUPAC Name N-phenyl-N-[(2,4,6-trinitrophenyl)-λ^{2}-azanyl]aniline
SMILES [O-][N+](=O)c1cc(c([N]N(c2ccccc2)c3ccccc3)c(c1)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H12N5O6/c24-21(25)15-11-16(22(26)27)18(17(12-15)23(28)29)19-20(13-7-3-1-4-8-13)14-9-5-2-6-10-14/h1-12H
Formula Weight 394.0788
Chemical Name or Material 2,2-Diphenyl-1-picrylhydrazyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.