missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dithiosalicylic acid, 98%

Product Code. 10254620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10254620 100 g 100g
10011050 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10254620 Supplier Thermo Scientific Acros Supplier No. 117561000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 119-80-2 | C14H10O4S2 | 306.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-80-2
Molecular Formula C14H10O4S2
Molecular Weight (g/mol) 306.34
MDL Number MFCD00002465
InChI Key LBEMXJWGHIEXRA-UHFFFAOYSA-N
Synonym 2,2'-dithiodibenzoic acid, 2,2'-dithiosalicylic acid, benzoic acid, 2,2'-dithiobis, 2-2-carboxyphenyl disulfanyl benzoic acid, bis 2-carboxyphenyl disulfide, bis o-carboxylphenyl disulfide, 2,2-dithiosalicylic acid, 2,2'-dithiobisbenzoic acid, unii-58mgb5279d, 2,2'-dithiodi benzoic acid
PubChem CID 8409
IUPAC Name 2-[(2-carboxyphenyl)disulfanyl]benzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)SSC2=CC=CC=C2C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 287°C to 290°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula S2(C6H4CO2H)2
Quantity 100 g
Beilstein 10, 129
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in dichloromethane,ether,benzene and,acetone
Formula Weight 306.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2, 2'-Dithiosalicylic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.