missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Furil, 98%

Product Code. 11478336
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11478336

Brand: Thermo Scientific Alfa Aesar A15386.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 492-94-4 | C10H6O4 | 190.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 492-94-4
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.154
MDL Number MFCD00003241
InChI Key SXPUVBFQXJHYNS-UHFFFAOYSA-N
Synonym furil, alpha-furil, bipyromucyl, 2,2'-furil, bipryomucyl, di-2-furylglyoxal, difuranylglyoxal, ethanedione, di-2-furanyl, di-2-furanylethanedione, a-furil
PubChem CID 68119
IUPAC Name 1,2-bis(furan-2-yl)ethane-1,2-dione
SMILES C1=COC(=C1)C(=O)C(=O)C2=CC=CO2

Specifications

Melting Point 162°C to 165°C
Quantity 25 g
Beilstein 383882
Formula Weight 190.15
Percent Purity 98%
Chemical Name or Material 2,2'-Furil

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.