missing translation for 'onlineSavingsMsg'
Learn More

[2-(2-Furyl)phenyl]methanol, ≥97%, Thermo Scientific™

Product Code. 10387404
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10387404 1 g 1g
10664215 250 mg 250mg
10032084 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10387404 Supplier Thermo Scientific Maybridge Supplier No. CC46209DA
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 139697-88-4
Molecular Formula C11H10O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD06203101
InChI Key NBLMKRIYULDNBF-UHFFFAOYSA-N
Synonym 2-2-furyl phenyl methanol, 2-furan-2-yl phenyl methanol, 2-fur-2-yl benzyl alcohol, 2-2-furyl benzenemethanol, 2-fur-2-yl phenyl methanol, benzenemethanol,2-2-furanyl, 2-2-furyl phenyl methan-1-ol
PubChem CID 10855985
IUPAC Name [2-(furan-2-yl)phenyl]methanol
SMILES OCC1=CC=CC=C1C1=CC=CO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 174.2
Percent Purity 97%
Chemical Name or Material [2-(2-Furyl)phenyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.