missing translation for 'onlineSavingsMsg'
Learn More

[2-(2-Furyl)phenyl]methylamine, ≥97%, Thermo Scientific™

Product Code. 10728275
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Dimensione della confezione
10728275 1 g Amber Glass Bottle 1g
10082414 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 10728275 Fornitore Thermo Scientific Maybridge N. del fornitore CC46213DA
 Restricted Item
Estimated Shipment: 10-12-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Identificatori chimici

CAS 771573-25-2
Molecular Formula C11H11NO
Molecular Weight (g/mol) 173.215
MDL Number MFCD06213664
InChI Key KUTREPQQCILHIM-UHFFFAOYSA-N
Synonym 2-2-furyl phenyl methylamine, 2-furan-2-yl phenyl methanamine, 1-2-furan-2-yl phenyl methanamine, 2-furan-2-yl-benzylamine
PubChem CID 18525791
IUPAC Name [2-(furan-2-yl)phenyl]methanamine
SMILES C1=CC=C(C(=C1)CN)C2=CC=CO2

Specifica

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 173.21
Percent Purity 97%
Chemical Name or Material [2-(2-Furyl)phenyl]methylamine
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.