missing translation for 'onlineSavingsMsg'
Learn More

[2-(2-Morpholinoethoxy)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10730544
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10730544 1 g Amber Glass Bottle 1g
10720544 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10730544 Supplier Thermo Scientific Maybridge Supplier No. CC41909DA
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 106276-04-4
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.299
MDL Number MFCD07772843
InChI Key MJDMCSJTOOAYPS-UHFFFAOYSA-N
Synonym 2-2-morpholinoethoxy phenyl methanol, 2-2-morpholin-4-yl ethoxy phenyl methanol, 2-2-morpholin-4-ylethoxy phenyl methanol, 2-2-morpholin-4-yl-ethoxy-phenyl-methanol, 2-2-morpholin-4-ylethoxy phenyl methan-1-ol
PubChem CID 7164615
IUPAC Name [2-(2-morpholin-4-ylethoxy)phenyl]methanol
SMILES C1COCCN1CCOC2=CC=CC=C2CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 237.3
Percent Purity 97%
Chemical Name or Material [2-(2-Morpholinoethoxy)phenyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.