missing translation for 'onlineSavingsMsg'
Learn More

2-(2-n-Butoxyethoxy)ethyl acetate, 98%

Product Code. 11463077
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mL
1000 mL
missing translation for 'unitSize'
1000mL
250mL
This item is not returnable. View return policy

Product Code. 11463077

missing translation for 'mfr': Thermo Scientific Alfa Aesar A16065.AU

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 124-17-4 | C10H20O4 | 204.266 g/mol

2-(2-n-Butoxyethoxy)ethyl acetate is widely used in high-grade paints for car, TV, freezer and plane. It is used as a solvent for enhancement of coating's characters in the coating industry. It is also used as a cleaning and furnishing care product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-n-Butoxyethoxy)ethyl acetate is widely used in high-grade paints for car, TV, freezer and plane. It is used as a solvent for enhancement of coating′s characters in the coating industry. It is also used as a cleaning and furnishing care product.

Solubility
Miscible with water, acetone, ethyl ether, ethanol and most organic liquids.

Notes
Light sensitive and heat sensitive. Incompatible with strong oxidizing agents, acids and strong bases. Keep the container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-17-4
Molecular Formula C10H20O4
Molecular Weight (g/mol) 204.266
MDL Number MFCD00009458
InChI Key VXQBJTKSVGFQOL-UHFFFAOYSA-N
Synonym 2-2-butoxyethoxy ethyl acetate, butoxyethoxyethyl acetate, diethylene glycol monobutyl ether acetate, butyl diglycol acetate, butyl carbitol acetate, ektasolve db acetate, 2-2-butoxyethoxy ethanol acetate, butyl diethylene glycol acetate, butylkarbitolacetat, diglycol monobutyl ether acetate
PubChem CID 31288
IUPAC Name 2-(2-butoxyethoxy)ethyl acetate
SMILES CCCCOCCOCCOC(=O)C

Specifications

Melting Point °C to 32°C
Density 0.977
Boiling Point 237°C to 241°C
Flash Point 115°C (239°F)
Assay Percent Range 98%
Refractive Index 1.426
Quantity 1000 mL
Beilstein 1771533
Solubility Information Miscible with water,acetone,ethyl ether,ethanol and most organic liquids.
Formula Weight 204.27
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-(2-n-Butoxyethoxy)ethyl Acetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.