missing translation for 'onlineSavingsMsg'
Learn More

2-[(2-Phenoxyethyl)thio]acetic acid, 97%, Thermo Scientific™

Product Code. 10039391
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10039391 1 g 1g
10540922 10 g 10g
10052074 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10039391 Supplier Thermo Scientific Maybridge Supplier No. KM06649DA
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 75434-70-7
Molecular Formula C10H12O3S
Molecular Weight (g/mol) 212.263
MDL Number MFCD00085152
InChI Key SHWKIHWUFNTMIB-UHFFFAOYSA-N
Synonym 2-2-phenoxyethyl thio acetic acid, 2-2-phenoxyethyl sulfanyl acetic acid, 2-phenoxyethyl sulfanyl acetic acid, maybridge1_005030, 2-2-phenoxyethylthio acetic acid, 2-phenoxy-ethylsulfanyl acetic acid, 2-phenoxy-ethylsulfanyl-acetic acid, 2-2-phenoxyethylsulfanyl acetic acid
PubChem CID 2779087
IUPAC Name 2-(2-phenoxyethylsulfanyl)acetic acid
SMILES C1=CC=C(C=C1)OCCSCC(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula C6H5O(CH2)2SCH2CO2H
Quantity 1 g
Formula Weight 212.27
Percent Purity 97%
Chemical Name or Material 2-[(2-Phenoxyethyl)thio]acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.