missing translation for 'onlineSavingsMsg'
Learn More

2-[3-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-oxopropyl]benzoic Acid Methyl Ester, TRC

Product Code. 30362261
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362261 25 mg 25mg
30366932 10 mg 10mg
30364620 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30362261 Supplier Toronto Research Chemicals Supplier No. TRCC38001025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149968-11-6
Molecular Formula C28 H22 Cl N O3
Molecular Weight (g/mol) 455.93
Synonym 2-[3-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-oxopropyl]benzoic Acid Methyl Ester, Benzoic acid, 2-[3-[3-[2-(7-chloro-2-quinolinyl)ethenyl]phenyl]-3-oxopropyl]-, methyl ester, (E)-, Methyl (E)-2-[3-[3-[2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-oxopropyl]benzoate, Methyl (E)-2-[3-[3-[2-(7-chloro-2-quinolinyl)ethenyl]phenyl]-3-oxopropyl]benzoate
IUPAC Name methyl 2-[3-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-oxopropyl]benzoate
SMILES COC(=O)c1ccccc1CCC(=O)c2cccc(\C=C\c3ccc4ccc(Cl)cc4n3)c2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C28H22ClNO3/c1-33-28(32)25-8-3-2-6-20(25)12-16-27(31)22-7-4-5-19(17-22)9-14-24-15-11-21-10-13-23(29)18-26(21)30-24/h2-11,13-15,17-18H,12,16H2,1H3/b14-9+
Formula Weight 455.1288
Chemical Name or Material 2-[3-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-oxopropyl]benzoic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.