missing translation for 'onlineSavingsMsg'
Learn More

2',3,3',4,4',5,5',6,6'-Nonabromo-[1,1'-biphenyl]-2-amine Hydrogen Sulfate Salt, TRC

Product Code. 30478123
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478123 10 mg 10mg
30475961 5 mg 5mg
30472055 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30478123 Supplier Toronto Research Chemicals Supplier No. TRCN64912010MG
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2',3,3',4,4',5,5',6,6'-Nonabromo-[1,1'-biphenyl]-2-amine Hydrogen Sulfate Salt
Analyte or Component Names 2',3,3',4,4',5,5',6,6'-Nonabromo-[1,1'-biphenyl]-2-amine Hydrogen Sulfate Salt
Shipping Condition Room Temperature
Molecular Formula C12 H2 Br9 N . H2 S O4
InChI Formula InChI=1S/C12H2Br9N.H2O4S/c13-3-1(4(14)7(17)9(19)6(3)16)2-5(15)8(18)10(20)11(21)12(2)22;1-5(2,3)4/h22H2;(H2,1,2,3,4)
Solution Type Neat
SMILES Nc1c(Br)c(Br)c(Br)c(Br)c1c2c(Br)c(Br)c(Br)c(Br)c2Br.OS(=O)(=O)O
IUPAC Name sulfuric acid;2,3,4,5-tetrabromo-6-(2,3,4,5,6-pentabromophenyl)aniline
Industry Type Pharmaceutical
Molecular Weight (g/mol) 977.365
Quantity 10 mg
Formula Weight 968.251
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.