missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentabromotoluene, TRC

Product Code. 17921045
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17921045 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17921045 Supplier Toronto Research Chemicals Supplier No. TRCP23788010G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 87-83-2
Molecular Formula C7 H3 Br5
Quantity 10 g
Synonym Pentabromotoluene, Benzene, pentabromomethyl- (9CI), Toluene, 2,3,4,5,6-pentabromo- (6CI,7CI,8CI), 1,2,3,4,5-Pentabromo-6-methylbenzene, 2,3,4,5,6-Pentabromomethylbenzene, 2,3,4,5,6-Pentabromotoluene, Flammex 5BT, PBT, PBT (flame retardant), Pentabromomethylbenzene, Pentabromotoluene, Benzene, pentabromomethyl- (9CI), Toluene, 2,3,4,5,6-pentabromo- (6CI,7CI,8CI), 1,2,3,4,5-Pentabromo-6-methylbenzene, 2,3,4,5,6-Pentabromomethylbenzene, 2,3,4,5,6-Pentabromotoluene, Flammex 5BT, PBT, PBT (flame retardant), Pentabromomethylbenzene, Pentabromotoluene
InChI Formula InChI=1S/C7H3Br5/c1-2-3(8)5(10)7(12)6(11)4(2)9/h1H3
SMILES Cc1c(Br)c(Br)c(Br)c(Br)c1Br
IUPAC Name 1,2,3,4,5-pentabromo-6-methylbenzene
Molecular Weight (g/mol) 486.62
Formula Weight 481.6152
Chemical Name or Material 2,3,4,5,6-Pentabromotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.