missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluoroaniline, 97%

Product Code. 10081560 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
250 g
Packaging:
Glass Bottle
Unit Size:
10g
250g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10081560 10 g Glass Bottle 10g
10247333 5 g Glass Bottle 5g
10006113 50 g Glass Bottle 50g
10239223 250 g Glass Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10081560 Supplier Thermo Scientific Acros Supplier No. 147080100
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 771-60-8 | C6H2F5N | 183.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-60-8
Molecular Formula C6H2F5N
Molecular Weight (g/mol) 183.08
MDL Number MFCD00007643
InChI Key NOXLGCOSAFGMDV-UHFFFAOYSA-N
Synonym pentafluoroaniline, pentafluorophenylamine, aminopentafluorobenzene, benzenamine, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluorobenzenamine, aniline, 2,3,4,5,6-pentafluoro, unii-ms06il3y2q, ms06il3y2q, 2,3,4,5,6-pentafluorophenylamine, pubchem1546
PubChem CID 13040
IUPAC Name 2,3,4,5,6-pentafluoroaniline
SMILES NC1=C(F)C(F)=C(F)C(F)=C1F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 33°C to 38°C
Color Salmon to White
Density 73g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula C6F5NH2
Quantity 10 g
Formula Weight 183.08
Percent Purity ≥96.0%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 2,3,4,5,6-Pentafluoroaniline
Show More Show Less
compliance-icons
Product Identifier
  • 2,3,4,5,6-Pentafluoroaniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.