missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorobenzyl bromide, 97%

Product Code. 11474713
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474713 100 g 100g
11454713 5 g 5g
11464713 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474713 Supplier Thermo Scientific Alfa Aesar Supplier No. A11623.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1765-40-8 | C7H2BrF5 | 260.989 g/mol

2,3,4,5,6-Pentafluorobenzyl bromide is used as a reagent for derivatization of alcohols, carboxylic acids, sulfonamides for GC and polyfunctional thiols. It is used for the preparation of pentafluorobenzyl esters of organic acids for determination by capillary and GC. It is also used to derivatize N-7-substituted guanine adducts of DNA for determination by GC-electron capture-MS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4,5,6-Pentafluorobenzyl bromide is used as a reagent for derivatization of alcohols, carboxylic acids, sulfonamides for GC and polyfunctional thiols. It is used for the preparation of pentafluorobenzyl esters of organic acids for determination by capillary and GC. It is also used to derivatize N-7-substituted guanine adducts of DNA for determination by GC-electron capture-MS.

Solubility
Hydrolyzes in water.

Notes
Store in a cool, dry, well-ventilated area. Incompatible with bases and oxidizing agents. Store at 4°C. Protected from moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1765-40-8
Molecular Formula C7H2BrF5
Molecular Weight (g/mol) 260.989
MDL Number MFCD00000299
InChI Key XDEPVFFKOVDUNO-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorobenzyl bromide, pentafluorobenzyl bromide, 1-bromomethyl-2,3,4,5,6-pentafluorobenzene, 1-bromomethylpentafluorobenzene, alpha-bromo-2,3,4,5,6-pentafluorotoluene, bromomethyl pentafluorobenzene, benzene, bromomethyl pentafluoro, 2,3,4,5,6-pentafluorobenzylbromide, pfbbr, pentafluorobenzylbromide
PubChem CID 74484
IUPAC Name 1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene
SMILES C(C1=C(C(=C(C(=C1F)F)F)F)F)Br

Specifications

Melting Point 18°C to 20°C
Density 1.844
Boiling Point 174°C to 175°C
Flash Point 83°C (181°F)
Refractive Index 1.472
Quantity 100 g
UN Number UN3265
Beilstein 647808
Solubility Information Hydrolyzes in water.
Formula Weight 261
Percent Purity ≥98%
Chemical Name or Material 2,3,4,5,6-Pentafluorobenzyl bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.