missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorostyrene, 97%, stabilized

Product Code. 10523171 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10523171 25 g Glass bottle 25g
10072570 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10523171 Supplier Thermo Scientific Acros Supplier No. 188470250
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 653-34-9 | C8H3F5 | 194.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 653-34-9 , 123-31-9
Molecular Formula C8H3F5
Molecular Weight (g/mol) 194.1
MDL Number MFCD00000300
InChI Key LVJZCPNIJXVIAT-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorostyrene, pentafluorostyrene, benzene, ethenylpentafluoro, 1,2,3,4,5-pentafluoro-6-vinyl-benzene, 2',3',4',5',6'-pentafluorostyrene, 2-2,3,4,5,6-pentafluorophenyl ethyl, vinylpentafluorobenzene, acmc-1avmt, styrene, 2,3,4,5,6-pentafluoro, 1,2,3,4,5-pentafluoro-6-vinylbenzene
PubChem CID 69556
IUPAC Name 1-ethenyl-2,3,4,5,6-pentafluorobenzene
SMILES C=CC1=C(C(=C(C(=C1F)F)F)F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4060g/mL
Boiling Point 62.0°C to 63.0°C (50.0 mmHg)
Flash Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6F5CH=CH2
Refractive Index 1.4445 to 1.4465
Quantity 25 g
Specific Gravity 1.406
Stabilizer 0.01% tert.-butylcatechol
Formula Weight 194.1
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2,3,4,5,6-Pentafluorostyrene, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2,3,4,5,6-Pentafluorostyrene, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.