missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorostyrene, 98%, stab. with 250ppm 4-tert-butylcatechol

Product Code. 11492858
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492858 5 g 5g
11402868 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11492858 Supplier Thermo Scientific Alfa Aesar Supplier No. A16833.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 653-34-9 | C8H3F5 | 194.104 g/mol

2,3,4,5,6-Pentafluorostyrene is used in the synthesis of optically active copolymers having low surface energies by utilizing beta-pinene as another monomer. It is also used in the synthesis of fluorinated and photo crosslinkable liquid prepolymers, which is used for flexible optical waveguides. Polymeric fluorinated nano particles prepared using 2,3,4,5,6-Pentafluorostyrene and styrene is explored in magnetic resonance image (MRI) applications, since it has larger structural design potential compared to traditional systems like emulsions and solutions of smaller molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4,5,6-Pentafluorostyrene is used in the synthesis of optically active copolymers having low surface energies by utilizing beta-pinene as another monomer. It is also used in the synthesis of fluorinated and photo crosslinkable liquid prepolymers, which is used for flexible optical waveguides. Polymeric fluorinated nano particles prepared using 2,3,4,5,6-Pentafluorostyrene and styrene is explored in magnetic resonance image (MRI) applications, since it has larger structural design potential compared to traditional systems like emulsions and solutions of smaller molecules.

Solubility
Immiscible with water.

Notes
Store in a cool and well ventilated area. Keep the container tightly closed, away from sources of ignition and direct sunlight. Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 653-34-9
Molecular Formula C8H3F5
Molecular Weight (g/mol) 194.104
MDL Number MFCD00000300
InChI Key LVJZCPNIJXVIAT-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorostyrene, pentafluorostyrene, benzene, ethenylpentafluoro, 1,2,3,4,5-pentafluoro-6-vinyl-benzene, 2',3',4',5',6'-pentafluorostyrene, 2-2,3,4,5,6-pentafluorophenyl ethyl, vinylpentafluorobenzene, acmc-1avmt, styrene, 2,3,4,5,6-pentafluoro, 1,2,3,4,5-pentafluoro-6-vinylbenzene
PubChem CID 69556
IUPAC Name 1-ethenyl-2,3,4,5,6-pentafluorobenzene
SMILES C=CC1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Density 1.406
Boiling Point 139°C to 140°C
Flash Point 34°C (93°F)
Odor Characteristic
Refractive Index 1.446
Quantity 5 g
UN Number UN1993
Beilstein 1874390
Solubility Information Immiscible with water.
Formula Weight 194.1
Percent Purity 98%
Chemical Name or Material 2,3,4,5,6-Pentafluorostyrene, stabilized with 250ppm 4-tert-butylcatechol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.