missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrafluoroaniline, 98%

Product Code. 15417958
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15417958 1 g 1g
15427958 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417958 Supplier Thermo Scientific Alfa Aesar Supplier No. H61417.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5580-80-3 | C6H3F4N | 165.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5580-80-3
Molecular Formula C6H3F4N
Molecular Weight (g/mol) 165.091
MDL Number MFCD00025153
InChI Key BEECAQIHCYTZHC-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluoro-phenylamine, benzenamine, 2,3,4,5-tetrafluoro, aniline, 2,3,4,5-tetrafluoro, benzenamine, tetrafluoro, aniline, 2,3,4,5-tetrafluoro-8ci, pubchem1547, acmc-1ajyg, 2,4,5-tetrafluoroaniline, ksc273e5j, aniline,3,4,5-tetrafluoro
PubChem CID 21757
IUPAC Name 2,3,4,5-tetrafluoroaniline
SMILES C1=C(C(=C(C(=C1F)F)F)F)N

Specifications

Melting Point 27°C to 29°C
Color White to Brown
Boiling Point 78°C
Flash Point 79°C (174°F)
Quantity 1 g
UN Number UN2811
Formula Weight 165.09
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3,4,5-Tetrafluoroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.