missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrafluorobenzoyl chloride, 98%, Thermo Scientific™

Product Code. 11410508
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410508 25 g 25g
11490498 1 g 1g
11400508 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410508 Supplier Thermo Scientific Alfa Aesar Supplier No. B24644.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94695-48-4
Molecular Formula C7HClF4O
Molecular Weight (g/mol) 212.53
MDL Number MFCD00075164
InChI Key XWCKIXLTBNGIHV-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluorobenzoylchloride, tetrafluorobenzoyl chloride, 2,3,4,5-tetrafluoro benzoyl chloride, benzoyl chloride, 2,3,4,5-tetrafluoro, 2,3,4,5-tetrafluorbenzoylchlorid, 2,3,4,5-tetrafluoro-benzoyl chloride, 2,3,4,5-tetrafluorobenzoic acid chloride, 2,3,4,5-tetrafluorobezoyl chloride, benzoyl chloride, 2,3,4,5-tetrafluoro-9ci, 2,3,4,5-tetrafluorobezoylchloride
PubChem CID 2733689
IUPAC Name 2,3,4,5-tetrafluorobenzoyl chloride
SMILES FC1=CC(C(Cl)=O)=C(F)C(F)=C1F

Specifications

Density 1.58
Boiling Point 65°C to 66°C (10 mmHg)
Flash Point 90°C (194°F)
Refractive Index 1.4787
Quantity 25 g
UN Number UN3265
Beilstein 5266860
Sensitivity Moisture sensitive
Formula Weight 212.53
Percent Purity 98%
Chemical Name or Material 2,3,4,5-Tetrafluorobenzoyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.