missing translation for 'onlineSavingsMsg'
Learn More

2-(3,4,5-Trifluorophenyl)ethanol, 96%, Thermo Scientific™

Product Code. 15495557
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495557 1 g 1g
15405567 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495557 Supplier Thermo Scientific Alfa Aesar Supplier No. H32800.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886761-77-9
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.138
MDL Number MFCD06660355
InChI Key VGWRUVISALLMOW-UHFFFAOYSA-N
Synonym 2-3,4,5-trifluorophenyl ethanol, 2-3,4,5-trifluorophenyl ethan-1-ol, benzeneethanol, 3,4,5-trifluoro, 3,4,5-trifluorophenethyl alcohol
PubChem CID 17750838
IUPAC Name 2-(3,4,5-trifluorophenyl)ethanol
SMILES C1=C(C=C(C(=C1F)F)F)CCO

Specifications

Refractive Index 1.4732
Quantity 1 g
Formula Weight 176.14
Percent Purity 96%
Chemical Name or Material 2-(3,4,5-Trifluorophenyl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.