missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trimethoxybenzeneboronic acid, 98%

Product Code. 11319825
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319825 1 g 1g
11329825 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319825 Supplier Thermo Scientific Alfa Aesar Supplier No. L19838.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118062-05-8 | C9H13BO5 | 212.008 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118062-05-8
Molecular Formula C9H13BO5
Molecular Weight (g/mol) 212.008
MDL Number MFCD01318183
InChI Key LDQLSQRFSNMANA-UHFFFAOYSA-N
Synonym 2,3,4-trimethoxyphenyl boronic acid, 2,3,4-trimethoxybenzeneboronic acid, 2,3,4-trimethoxyphenyl boranediol, 2,3,4-trimethoxylphenylboronic acid, boronic acid, 2,3,4-trimethoxyphenyl, pubchem11893, acmc-2099vt, trimethoxyphenyl boronic acid, 2,3,4-trimethoxyphenylboronsaure, ablock ab-12-9176
PubChem CID 2773582
IUPAC Name (2,3,4-trimethoxyphenyl)boronic acid
SMILES B(C1=C(C(=C(C=C1)OC)OC)OC)(O)O

Specifications

Melting Point 74°C to 79°C
Quantity 1 g
Beilstein 4185924
Formula Weight 212.01
Percent Purity 98%
Chemical Name or Material 2,3,4-Trimethoxybenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.