missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5,6-Tetrafluoro-4-(methoxymethyl)benzyl alcohol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 4-Methoxymethyl-2,3,5,6-tetrafluorobenzyl Alcohol |
| CAS | 83282-91-1 |
| Analyte or Component Names | 4-Methoxymethyl-2,3,5,6-tetrafluorobenzyl Alcohol |
| Shipping Condition | +20°C |
| Molecular Formula | C9H8F4O2 |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | 2,3,5,6-Tetrafluoro-4-(methoxymethyl)-benzenemethanol, 2,3,5,6-Tetrafluoro-4-(methoxymethyl)benzyl Alcohol |
| SMILES | COCc1c(F)c(F)c(CO)c(F)c1F |
| Molecular Weight (g/mol) | 224.15 |
| Quantity | 250 mg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?