missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluoroanisole, 97%

Product Code. 11485677
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485677 25 g 25g
11475677 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485677 Supplier Thermo Scientific Alfa Aesar Supplier No. B24275.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2324-98-3 | C7H4F4O | 180.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2324-98-3
Molecular Formula C7H4F4O
Molecular Weight (g/mol) 180.10
MDL Number MFCD00000311
InChI Key AXCOCGJDERQVDK-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluoroanisole, 2,3,5,6-tetrafluoromethoxy benzene, anisole, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluoromethoxybenzene, methyl 2,3,5,6-tetrafluorophenyl ether, benzene, 1,2,4,5-tetrafluoro-3-methoxy, 1,2,4,5-tetrafluoro-3-methoxy-benzene, pubchem10114, acmc-1clho, 2,3,5,6-tetrafluoro-anisole
PubChem CID 75351
IUPAC Name 1,2,4,5-tetrafluoro-3-methoxybenzene
SMILES COC1=C(F)C(F)=CC(F)=C1F

Specifications

Density 1.293
Boiling Point 144°C to 145°C
Flash Point 41°C (107°F)
Refractive Index 1.428
Quantity 25 g
UN Number UN1993
Formula Weight 180.1
Percent Purity 97%
Chemical Name or Material 2,3,5,6-Tetrafluoroanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.