missing translation for 'onlineSavingsMsg'
Learn More

2-[3,5-di(tert-butyl)-4-hydroxyphenyl]acetic acid, Thermo Scientific™

Product Code. 10326984
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10326984 25 g 25g
10210563 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10326984 Supplier Thermo Scientific Maybridge Supplier No. BTB09885EB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1611-03-6
Molecular Formula C16H24O3
Molecular Weight (g/mol) 264.365
InChI Key QLMGIWHWWWXXME-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-4-hydroxyphenylacetic acid, 3,5-di-tert-butyl-4-hydroxyphenyl acetic acid, 2-3,5-di tert-butyl-4-hydroxyphenyl acetic acid, 2-3,5-di-tert-butyl-4-hydroxyphenyl acetic acid, benzeneacetic acid,3,5-bis 1,1-dimethylethyl-4-hydroxy, acmc-1bvft, 4-hydroxy-3,5-di-t-butylphenylacetic acid, 3,5-di-t-butyl-4-hydroxyphenyl-acetic acid, 4-hydroxy-3,5-di-t-butyl-phenylacetic acid, 3,5-di-tert-butyl-4-hydroxybenzeneacetic acid
PubChem CID 2775110
IUPAC Name 2-(3,5-ditert-butyl-4-hydroxyphenyl)acetic acid
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CC(=O)O

Specifications

Packaging Amber glass bottle
Quantity 25 g
Formula Weight 264.36
Percent Purity 97%
Chemical Name or Material 2-[3, 5-di(tert-butyl)-4-hydroxyphenyl]acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.