missing translation for 'onlineSavingsMsg'
Learn More

2-(3,5-Dimethyl-1H-pyrazol-4-yl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10602633
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10602633

Brand: Thermo Scientific Maybridge CC15701CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 321309-43-7
Molecular Formula C12H12N2O2
Molecular Weight (g/mol) 216.24
MDL Number MFCD02682016
InChI Key JNHKHFADRGJMJS-UHFFFAOYSA-N
Synonym 2-3,5-dimethyl-1h-pyrazol-4-yl benzoic acid, 4-2-carboxyphenyl-3,5-dimethylpyrazole, benzoic acid,2-3,5-dimethyl-1h-pyrazol-4-yl, 2-3,5-dimethylpyrazol-4-yl benzoic acid, 2-3,5-dimethyl-1h-pyrazol-4-yl-benzoic acid
PubChem CID 2776446
IUPAC Name 2-(3,5-dimethyl-1H-pyrazol-4-yl)benzoic acid
SMILES CC1=C(C(C)=NN1)C1=CC=CC=C1C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 216.24
Percent Purity 97%
Chemical Name or Material 2-(3, 5-Dimethyl-1H-pyrazol-4-yl)benzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.