missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trimethylphenol, 98+%

Product Code. p-7080976
Click to view available options
Quantity:
25 g
100 g
1000 g
Unit Size:
1000g
100g
25g
This item is not returnable. View return policy

Product Code. 11494110

Brand: Thermo Scientific Alfa Aesar A10745.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 697-82-5 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 697-82-5
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002228
InChI Key OGRAOKJKVGDSFR-UHFFFAOYSA-N
Synonym isopseudocumenol, phenol, 2,3,5-trimethyl, 1-hydroxy-2,3,5-trimethylbenzene, unii-1s1061zbq1, phenol, 2,3,5 or 3,4,5-trimethyl, pubchem2249, 2,3,5-trimethyphenol, phenol,3,5-trimethyl, 2,3,5-trimethyl-phenol, 6-hydroxypseudocumene
PubChem CID 12769
ChEBI CHEBI:38570
IUPAC Name 2,3,5-trimethylphenol
SMILES CC1=CC(=C(C(=C1)O)C)C

Specifications

Melting Point 92°C to 95°C
Boiling Point 230°C to 231°C
Odor Phenol-like
Quantity 25 g
UN Number UN2430
Beilstein 2042210
Formula Weight 136.2
Percent Purity ≥98%
Chemical Name or Material 2,3,5-Trimethylphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.