missing translation for 'onlineSavingsMsg'
Learn More

2,3,6-Trifluorobenzaldehyde, 97%, Thermo Scientific™

Product Code. 10250033 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10250033 5 g Glass bottle 5g
10077052 1 g Glass bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10250033

Brand: Thermo Scientific Acros 384180050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 104451-70-9
Molecular Formula C7H3F3O
Molecular Weight (g/mol) 160.10
MDL Number MFCD00061195
InChI Key XSBAHBVACIKRTG-UHFFFAOYSA-N
Synonym benzaldehyde, 2,3,6-trifluoro, 2,3,6-trifluoro-benzaldehyde, pubchem1467, acmc-1bqm6, 2,3,6-trifluorobenzoaldehyde, ksc494g2t, 2,3,6-trifluoro benzaldehyde, timtec-bb sbb003970, attercop-chm at111497
PubChem CID 517845
IUPAC Name 2,3,6-trifluorobenzaldehyde
SMILES FC1=CC=C(F)C(C=O)=C1F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Boiling Point 68°C to 70°C (10.0 mmHg)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula F3C6H2CHO
Refractive Index 1.4870 to 1.489
Quantity 5 g
Formula Weight 160.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 3, 6-Trifluorobenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2,3,6-Trifluorobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.