missing translation for 'onlineSavingsMsg'
Learn More

2,3,6-Trifluorophenylacetic acid, 98%

Product Code. 10423464 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10423464 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10423464 Supplier Thermo Scientific Acros Supplier No. 426030010
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
This item is not returnable. View return policy

CAS: 114152-23-7 | C8H5F3O2 | 190.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 114152-23-7
CAS Min % 97.5
CAS Max % 100.0
Melting Point 111.0°C to 114.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H5F3O2
Linear Formula F3C6H2CH2COOH
MDL Number MFCD00061217
Quantity 1 g
Synonym 2,3,6-trifluorophenylacetic acid, 2-2,3,6-trifluorophenyl acetic acid, 2,3,6-trifluorophenyl acetic acid, benzeneacetic acid, 2,3,6-trifluoro, pubchem7293, acmc-1c5f9, rarechem al bo 0509, 2,3,6-trifluorophenylaceticacid, 2,3,6-trifluorophenylaceitc acid, 2,3,6-trifluorobenzeneacetic acid
InChI Key QRAZASHLGLHKEB-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1F)CC(=O)O)F)F
IUPAC Name 2-(2,3,6-trifluorophenyl)acetic acid
Molecular Weight (g/mol) 190.12
PubChem CID 2777948
Formula Weight 190.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3, 6-Trifluorophenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,3,6-Trifluorophenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.