missing translation for 'onlineSavingsMsg'
Learn More

2,3,6-Trifluoropyridine, 97%

Product Code. 11338926
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338926 1 g 1g
11348926 5 g 5g
11358926 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11338926 Supplier Thermo Scientific Alfa Aesar Supplier No. L19540.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3512-18-3 | C5H2F3N | 133.073 g/mol

2,3,6-Trifluoropyridine is used as an intermediate in organic syntheses and in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,6-Trifluoropyridine is used as an intermediate in organic syntheses and in pharmaceuticals.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3512-18-3
Molecular Formula C5H2F3N
Molecular Weight (g/mol) 133.073
MDL Number MFCD03001158
InChI Key HRLIANGJWPJFKW-UHFFFAOYSA-N
Synonym pyridine, 2,3,6-trifluoro, 2,3,6-trifluoro-pyridine, pubchem7955, acmc-1cs48, 2,3,6-tris fluoranyl pyridine
PubChem CID 2783288
IUPAC Name 2,3,6-trifluoropyridine
SMILES C1=CC(=NC(=C1F)F)F

Specifications

Density 1.402
Boiling Point 100°C to 102°C
Flash Point 30°C (86°F)
Refractive Index 1.42
Quantity 1 g
UN Number UN1993
Beilstein 1365146
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 133.07
Percent Purity 97%
Chemical Name or Material 2,3,6-Trifluoropyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.