missing translation for 'onlineSavingsMsg'
Learn More

2,3,6-Trimethylphenol, 95%

Product Code. 15446958
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446958 100 g 100g
15436958 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15446958 Supplier Thermo Scientific Alfa Aesar Supplier No. H60489.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2416-94-6 | C9H12O | 136.19 g/mol

2,3,6-Trimethylphenol is used as intermediate for synthetic vitamin E. It is used for manufacturing 2, 3, 5-Trimethylhydroquinone. It is used as intermediate for antioxidants and plastics. It is used as a comonomer for the modification of polyphenylene oxide resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,6-Trimethylphenol is used as intermediate for synthetic vitamin E. It is used for manufacturing 2, 3, 5-Trimethylhydroquinone. It is used as intermediate for antioxidants and plastics. It is used as a comonomer for the modification of polyphenylene oxide resins.

Solubility
Soluble in methanol.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2416-94-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00002229
InChI Key QQOMQLYQAXGHSU-UHFFFAOYSA-N
Synonym 3-hydroxypseudocumene, phenol, 2,3,6-trimethyl, 1-hydroxy-2,3,6-trimethylbenzene, 2,3,6-trimethyl-phenol, unii-05wkl2l5lj, 2,3,6-trimethyl phenol, 05wkl2l5lj, dsstox_cid_2187, dsstox_rid_76518, dsstox_gsid_22187
PubChem CID 17016
IUPAC Name 2,3,6-trimethylphenol
SMILES CC1=CC=C(C)C(O)=C1C

Specifications

Melting Point 59°C to 62°C
Density 0.94
Boiling Point 215°C
Flash Point 100°C (212°F)
Quantity 100 g
UN Number UN2430
Beilstein 2042207
Solubility Information Soluble in methanol.
Formula Weight 136.19
Percent Purity 95%
Chemical Name or Material 2,3,6-Trimethylphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.