missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromo-4-fluorophenyl)acetonitrile, 96%

Product Code. 15458808
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458808 5 g 5g
15468808 250 mg 250mg
15448808 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15458808 Supplier Thermo Scientific Alfa Aesar Supplier No. H62260.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 501420-63-9 | C8H5BrFN | 214.037 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501420-63-9
Molecular Formula C8H5BrFN
Molecular Weight (g/mol) 214.037
MDL Number MFCD08458154
InChI Key ORKCKZRBHXMWBO-UHFFFAOYSA-N
Synonym 2-3-bromo-4-fluorophenyl acetonitrile, 3-bromo-4-fluorophenylacetonitrile, 3-bromo-4-fluorobenzyl cyanide, benzeneacetonitrile, 3-bromo-4-fluoro, 3-bromo-4-fluorophenyl acetonitrile, 3-bromo-4-fluoro benzyl cyanide, 3-bromo-4-fluorobenzeneacetonitrile, 2-3-bromo-4-fluorophenyl ethanenitrile
PubChem CID 20113897
IUPAC Name 2-(3-bromo-4-fluorophenyl)acetonitrile
SMILES C1=CC(=C(C=C1CC#N)Br)F

Specifications

Quantity 5 g
UN Number UN3439
Formula Weight 214.03
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-(3-Bromo-4-fluorophenyl)acetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.