missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Bromomethyl)phenyl]thiophene, ≥97%, Thermo Scientific™

Product Code. 10536534
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10536534 250 mg 250mg
10528823 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10536534 Supplier Thermo Scientific Maybridge Supplier No. CC44508CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 85553-44-2
Molecular Formula C11H9BrS
Molecular Weight (g/mol) 253.157
MDL Number MFCD08435867
InChI Key JQQDEBDYUAKNGD-UHFFFAOYSA-N
Synonym 2-3-bromomethyl phenyl thiophene, 3-2-thienyl benzyl bromide, 2-3-bromomethyl phenyl-thiophene
PubChem CID 13150400
IUPAC Name 2-[3-(bromomethyl)phenyl]thiophene
SMILES C1=CC(=CC(=C1)CBr)C2=CC=CS2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 253.16
Percent Purity 97%
Chemical Name or Material 2-[3-(Bromomethyl)phenyl]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.