missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Bromomethyl)phenyl]thiophene, ≥97%, Thermo Scientific™

Product Code. p-3174844
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10536534

Brand: Thermo Scientific Maybridge CC44508CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 85553-44-2
Molecular Formula C11H9BrS
Molecular Weight (g/mol) 253.157
MDL Number MFCD08435867
InChI Key JQQDEBDYUAKNGD-UHFFFAOYSA-N
Synonym 2-3-bromomethyl phenyl thiophene, 3-2-thienyl benzyl bromide, 2-3-bromomethyl phenyl-thiophene
PubChem CID 13150400
IUPAC Name 2-[3-(bromomethyl)phenyl]thiophene
SMILES C1=CC(=CC(=C1)CBr)C2=CC=CS2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 253.16
Percent Purity 97%
Chemical Name or Material 2-[3-(Bromomethyl)phenyl]thiophene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.