missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromophenoxy)pyridine, 97%, Thermo Scientific™

Product Code. 10587684
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10587684 250 mg Amber Glass Bottle 250mg
10367264 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10587684 Supplier Thermo Scientific Maybridge Supplier No. CC50210CB
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 92545-83-0
Molecular Formula C11H8BrNO
Molecular Weight (g/mol) 250.10
MDL Number MFCD08741430
InChI Key CXEIVVAAUANDIA-UHFFFAOYSA-N
Synonym 2-3-bromophenoxy pyridine, 2-3-bromophenoxyl pyridine, pyridine, 2-3-bromophenoxy, 3-bromophenyl pyridin-2-yl ether, 3-bromo-1-2-pyridyloxy benzene, 1-bromo-3-pyridin-2-yl oxy benzene
PubChem CID 21616612
IUPAC Name 2-(3-bromophenoxy)pyridine
SMILES BrC1=CC=CC(OC2=CC=CC=N2)=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 250.09
Percent Purity 97%
Chemical Name or Material 2-(3-Bromophenoxy)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.